missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13446-1-AP-20UL
This item is not returnable.
View return policy
Description
The product of this gene is a microtubule-associated protein that is predomitly expressed in cells of epithelial origin. Microtubule-associated proteins are thought to be involved in microtubule dynamics, which is essential for cell polarization and differentiation. This protein has been shown to be able to stabilize microtubules, and may serve to modulate microtubule functions. Studies of the related mouse protein also suggested an essential role in microtubule function required for spermatogenesis. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| MAP7 | |
| Polyclonal | |
| Unconjugated | |
| MAP7 | |
| E MAP 115, EMAP115, Ensconsin, MAP 7, MAP7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17761, 293016, 9053 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O88735, Q14244 | |
| Map7 | |
| MAP7 Fusion Protein Ag4276 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction