missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MAP7 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5109935

Product Code. 17177863

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: AREQREKEER ERREQEELER QKREELAQRV AEERTTRREE ESRRLEAEQA REKEEQLQRQ AEERALRERE EAERAQRQKE EEARVREEAE RVRQEREKHF QREEQERLER KKRLEEIMKR TRRTEATDKK TSDQRNGDIA KGALTGGTEV SALPCTTNAP GNGKPVGSPH VVTSHQSKVT VESTPDLEKQ PNENGVSVQN ENFEEIINLP IGSKPSRLDV TNSESPEIPL NPILAFDDEG TLGPLPQVDG VQTQQTAEVI.

The product of this gene is a microtubule-associated protein that is predominantly expressed in cells of epithelial origin. Microtubule-associated proteins are thought to be involved in microtubule dynamics, which is essential for cell polarization and differentiation. This protein has been shown to be able to stabilize microtubules, and may serve to modulate microtubule functions. Studies of the related mouse protein also suggested an essential role in microtubule function required for spermatogenesis. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MAP7
Polyclonal
Unconjugated
MAP7
dJ325F22.2 (microtubule-associated protein 7 (EMAP115, E-MAP-115)); EMAP115; E-MAP-115; Ensconsin; epithelial microtubule-associated protein of 115 kDa; MAP7; MAP-7; microtubule associated protein 7; microtubule-associated protein 7; mshi; mste; Mtap7; R75000; ste
Rabbit
Affinity Chromatography
RUO
9053
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
3.35 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q14244
MAP7
Recombinant fusion protein containing a sequence corresponding to amino acids 490-749 of human MAP7 (NP_003971.1).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.