missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAP7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556235
This item is not returnable.
View return policy
Description
Immunogen sequence: PHLPGTPRPT SSLPPGSVKA APAQVRPPSP GNIRPVKREV KVEPEKKDPE KEPQKVANEP SLKGRAPLVK VEEATVEERT PAEPEVGPAA P Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 70%.
The product of this gene is a microtubule-associated protein that is predominantly expressed in cells of epithelial origin. Microtubule-associated proteins are thought to be involved in microtubule dynamics, which is essential for cell polarization and differentiation. This protein has been shown to be able to stabilize microtubules, and may serve to modulate microtubule functions. Studies of the related mouse protein also suggested an essential role in microtubule function required for spermatogenesis. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| MAP7 | |
| Polyclonal | |
| Unconjugated | |
| MAP7 | |
| dJ325F22.2 (microtubule-associated protein 7 (EMAP115, E-MAP-115)); EMAP115; E-MAP-115; Ensconsin; epithelial microtubule-associated protein of 115 kDa; MAP7; MAP-7; microtubule associated protein 7; microtubule-associated protein 7; mshi; mste; Mtap7; R75000; ste | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9053 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14244 | |
| MAP7 | |
| Recombinant protein corresponding to Human MAP7. Recombinant protein control fragment (Product #RP-94845). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction