missing translation for 'onlineSavingsMsg'
Learn More

MAP7 Antibody, Novus Biologicals™

Product Code. 18326548 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18326548 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18326548 Supplier Novus Biologicals Supplier No. H00009053B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

MAP7 Polyclonal antibody specifically detects MAP7 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP7
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation PBS (pH 7.4)
Gene Accession No. NP_003971.1
Gene Alias E-MAP-115dJ325F22.2 (microtubule-associated protein 7 (EMAP115, E-MAP-115)), ensconsin, Epithelial microtubule-associated protein of 115 kDa, MAP-7, microtubule-associated protein 7EMAP115
Host Species Mouse
Immunogen MAP7 (NP_003971.1, 1 a.a. - 749 a.a.) full-length human protein. MAELGAGGDGHRGGDGAVRSETAPDSYKVQDKKNASSRPASAISGQNNNHSGNKPDPPPVLRVDDRQRLARERREEREKQLAAREIVWLEREERARQHYEKHLEERKKRLEEQRQKEERRRAAVEEKRRQRLEEDKERHEAVVRRTMERSQKPKQKHNRWSWGGSLHGSPSIHSADPDRRSVSTMNLSKYVDPVISKRLSSSSATLLNSPDRARRLQLSPWESSVVNRLLTPTHSFLARSKSTAALSGEAASCSPIIMPYKAAHSRNSMDRPKLFVTPPEGSSRRRIIHGTASYKKERERENVLFLTSGTRRAVSPSNPKARQPARSRLWLPSKSLPHLPGTPRPTSSLPPGSVKAAPAQVRPPSPGNIRPVKREVKVEPEKKDPEKEPQKVANEPSLKGRAPLVKVEEATVEERTPAEPEVGPAAPAMAPAPASAPAPASAPAPAPVPTPAMVSAPSSTVNASASVKTSAGTTDPEEATRLLAEKRRLAREQREKEERERREQEELERQKREELAQRVAEERTTRREEESRRLEAEQAREKEEQLQRQAEERALREREEAERAQRQKEEEARVREEAERVRQEREKHFQREEQERLERKKRLEEIMKRTRRTEATDKKTSDQRNGDIAKGALTGGTEVSALPCTTNAPGNGKPVGSPHVVTSHQSKVTVESTPDLEKQPNENGVSVQNENFEEIINLPIGSKPSRLDVTNSESPEIPLNPILAFDDEGTLGPLPQVDGVQTQQTAEVI
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9053
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.