missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAP6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578399
This item is not returnable.
View return policy
Description
Positive Control: rat brain, mouse brain Predicted Reactivity: Human (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
STOP encodes a microtubule-associated protein. The encoded protein is a calmodulin-binding and calmodulin-regulated protein that is involved in microtubule stabilization. Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
| MAP6 | |
| Polyclonal | |
| Unconjugated | |
| Map6 | |
| 145-kDa STOP; 2810411E12Rik; CTD-2530H12.7; KIAA1878; LOW QUALITY PROTEIN: microtubule-associated protein 6; MAP6; MAP-6; MAP6-N; microtubule associated protein 6; microtubule-associated protein 6; microtubule-associated protein 6 homolog; Mtap6; N STOP; N-STOP; Stable tubule-only polypeptide; STOP; STOP145 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17760, 29457, 4135 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Western Blot | |
| 0.53 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q63560, Q7TSJ2, Q96JE9 | |
| Map6 | |
| Recombinant protein encompassing a sequence within the center region of human STOP. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction