Learn More
Thermo Scientific™ Human MAP4K3 (GLK), GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots; aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Recombinant human MAP4K3 (1-380) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Validated in LanthaScreen™ Kinase Binding Assay Protocols available for simple use and optimized performance Multiple pack sizes available for convenient use Related Product:LanthaScreen™ Eu Kinase Binding Assay Protein Form: Recombinant, Fragment The gene accession number for MAP4K3 (GLK) is NP_003609.2.
Specifications
Specifications
| Accession Number | Q8IVH8 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50mM tris HCl with 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, 150mM NaCl, 0.25mM DTT, 25% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 8491 |
| Molecular Weight (g/mol) | 69.6 kDa |
| Name | Human MAP4K3 (GLK), GST Tag |
| pH Range | 7.5 |
| Purification Method | Purified |
| Quantity | 100 μg |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.