missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP4K2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55244-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to MAP4K2.
The protein encoded by this gene is a member of the serine/threonine protein kinase family. Although this kinase is found in many tissues, its expression in lymphoid follicles is restricted to the cells of germinal centre, where it may participate in B-cell differentiation. This kinase can be activated by TNF-alpha, and has been shown to specifically activate MAP kinases. This kinase is also found to interact with TNF receptor-associated factor 2 (TRAF2), which is involved in the activation of MAP3K1/MEKK1.Specifications
| MAP4K2 | |
| Polyclonal | |
| Unconjugated | |
| MAP4K2 | |
| BL44, GC kinase, GCK, Germinal center kinase, MAP4K2, MEK kinase kinase 2, MEKKK 2, Rab8 interacting protein, RAB8IP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26412, 293694, 5871 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.47 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q12851, Q61161 | |
| MAP4K2 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto