Learn More
Invitrogen™ Human MAP4K2 (GCK), His Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length
Specifications
Specifications
| Accession Number | Q12851 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 5871 |
| Molecular Weight (g/mol) | 95.6 kDa |
| Name | Human MAP4K2 (GCK), His Tag |
| Quantity | 10 μg |
| Regulatory Status | RUO - research use only |
| Gene Alias | AI385662; B lymphocyte serine/threonine protein kinase; B lymphocyte serine/threonine-protein kinase; BL44; dZ150L22.2; GC kinase; GCK; germinal center kinase; germinal centre kinase (GC kinase); LOW QUALITY PROTEIN: mitogen-activated protein kinase kinase kinase kinase 2; MAP4K2; map4k2l; map4k2l-2; map4k3l; MAPK/ERK kinase kinase kinase 2; MEK kinase kinase 2; MEKKK 2; mitogen activated protein kinase kinase kinase kinase 2; mitogen-activated protein kinase kinase kinase kinase 2; mitogen-activated protein kinase kinase kinase kinase 2-like; mitogen-activated protein kinase kinase kinase kinase 3-like; Rab8 interacting protein; rab8-interacting protein; Rab8ip; si:dz150l22.2; si:dz263j20.1; Unknown (protein for MGC:136670); zgc:136670 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.