Learn More
Invitrogen™ Human MAP4K1 (HPK1), GST Tag Recombinant Protein
Human MAP4K1 (HPK1), GST Tag Recombinant Protein
Brand: Invitrogen™ PV6357
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MAP4K1 is a tissue-specific upstream activator of the MEKK/JNK/SAPK signaling pathway. MAP4K1 diminishes T cell receptor (TCR) signaling activity and T cell proliferation by phosphorylating the adaptor protein SLP-76.
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
65.0 kDa | |
MAP4K1 | |
Human | |
GST Tag | |
Serine/Threonine Kinase | |
Kinases & Inhibitors | |
Liquid |
1mg | |
aa 1-346 | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl pH 7.5, 150 mM NaCl, 0.25 mM DTT, 10 mM glutathione, 0.1 mM EDTA, 0.1 mM PMSF, 25% glycerol Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. |
|
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |