missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP4 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68271-1-IG-20UL
This item is not returnable.
View return policy
Description
The MAP4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MAP4. This antibody recognizes human antigen. The MAP4 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Specifications
| MAP4 | |
| Monoclonal | |
| 500 μg/mL | |
| DKFZp779A1753; MGC8617 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| 210-240 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| 1C1A6 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 4134 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction