missing translation for 'onlineSavingsMsg'
Learn More

MAP3K8/Tpl2/COT Antibody (OTI3D9), DyLight 594, Novus Biologicals™

Product Code. 18409267 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409267 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409267 Supplier Novus Biologicals Supplier No. NBP272589DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MAP3K8/Tpl2/COT Monoclonal antibody specifically detects MAP3K8/Tpl2/COT in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP3K8/Tpl2/COT
Applications Western Blot
Classification Monoclonal
Clone OTI3D9
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias Cancer Osaka thyroid oncogene, c-COT, cot (cancer Osaka thyroid) oncogene, COTMEKK8, EC 2.7.11.25, ESTF, ESTFLJ10486, Ewing sarcoma transformant, mitogen-activated protein kinase kinase kinase 8, Proto-oncogene c-Cot, proto-oncogene serine/threoine protein kinase, Serine/threonine-protein kinase cot, Tpl-2, TPL2, Tumor progression locus 2, tumor progression locus-2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 277-461 of human MAP3K8/Tpl2/COT (NP_005195) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 1326
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.