missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP3K8/Tpl2/COT Antibody (OTI3D9), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72589CL1
This item is not returnable.
View return policy
Description
MAP3K8/Tpl2/COT Monoclonal antibody specifically detects MAP3K8/Tpl2/COT in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| MAP3K8/Tpl2/COT | |
| Monoclonal | |
| CoraFluor 1 | |
| Cancer Osaka thyroid oncogene, c-COT, cot (cancer Osaka thyroid) oncogene, COTMEKK8, EC 2.7.11.25, ESTF, ESTFLJ10486, Ewing sarcoma transformant, mitogen-activated protein kinase kinase kinase 8, Proto-oncogene c-Cot, proto-oncogene serine/threoine protein kinase, Serine/threonine-protein kinase cot, Tpl-2, TPL2, Tumor progression locus 2, tumor progression locus-2 | |
| Human recombinant protein fragment corresponding to amino acids 277-461 of human MAP3K8(NP_005195) produced in E.coli. | |
| 0.1 mL | |
| Angiogenesis, Cancer, Protein Kinase | |
| 1326 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Western Blot | |
| OTI3D9 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction