missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MAP3K7/MAP3K7IP1 (TAK1-TAB1), His Tag Recombinant Protein
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Descripción
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length
MAP3K7 (TAK1) controls transcription regulation and apoptosis, and is implicated in inflammatory disease and rheumatoid arthritis. Fusion of MAP3K7 to MAP3K7IP1 (TAB1) activates MAP3K7. Originally identified as a TGF- beta-activated kinase, TAK1 interacts with TGF- beta-receptor and TRAF6 to modulate TGF- beta activation of JNK and p38. It has additional roles in activation of p38 and JNK through Wnt, BMP, and activin signaling pathways as well in response to thyroid hormone, osmotic stress, endothelin, and ephrine. TGF-beta-activated kinase 1 and MAP3K7-binding protein 1 (TAB1) is a regulator of the MAP kinase kinase kinase MAP3K7/TAK1, which is known to mediate various intracellular signaling pathways, such as those induced by TGF beta, interleukin 1, and WNT-1. TAB1 interacts and thus activates TAK1 kinase.
Especificaciones
Especificaciones
| Accession Number | O43318, Q15750 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 10454, 6885 |
| Molecular Weight (g/mol) | 45.8 kDa |
| Name | Human MAP3K7/MAP3K7IP1 (TAK1-TAB1), His Tag |
| Quantity | 100 μg |
| Regulatory Status | RUO - research use only |
| Gene Alias | TAB1; TAK1 |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido