missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP3K5, Rabbit anti-Human, SureLight 488, Clone: G4, Abnova™
Rabbit Monoclonal Antibody
Brand: Abnova MAB18795.100Reactions
This item is not returnable.
View return policy
Specifications
| MAP3K5 | |
| Monoclonal | |
| SureLight 488 | |
| In PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide). | |
| ASK1/MAPKKK5/MEKK5 | |
| Rabbit | |
| 100 Reactions | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| G4 | |
| Flow Cytometry (5 uL/106 cells) The optimal working dilution should be determined by the end user. | |
| mitogen-activated protein kinase kinase kinase 5 | |
| MAP3K5 | |
| A synthetic phosphopeptide corresponding to residues surrounding S83 of human MAP3K5. | |
| RUO | |
| 4217 | |
| Store at 4°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction