missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MAP3K5 (ASK1), GST Tag Recombinant Protein

Product Code. 11523547
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11523547

Brand: Invitrogen™ PV3809

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Full Length.

MAP3K5 (ASK1) is a serine/threonine kinase that is abundantly expressed in human heart and pancreas and are activated in response to various extracellular stimuli, including cytokines, growth factors and environmental stresses. A novel MAP kinase kinase kinase (MAPKKK) was identified and designated ASK1 (for apoptosis signal-regulating kinase 1) and MAPKKK5. ASK1 activates two different subgroups of MAPKK, MKK4 and MKK6, which in turn activates c-Jun N-terminal kinase (JNK) and p38 MAP kinase, respectively. ASK1/MAPKKK5 is activated by TNFR and Fas through the interaction with members of the TRAF family and Fas-associated protein Daxx. Overexpression of ASK1 induced apoptotic cell death and a catalytically inactive form of ASK1 inhibited TNF-a-induced apoptosis. ASK1 is expressed in variety of human and mouse tissues.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q99683
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 0.5mM EDTA, 50% glycerol, 150mM NaCl, 2mM DTT, 0.02% Triton X-100 and no preservative; pH 7.5
Gene ID (Entrez) 4217
Molecular Weight (g/mol) 182 kDa
Name Human MAP3K5 (ASK1), GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 7420452D20Rik; apoptosis signal regulating kinase 1; apoptosis signal-regulating kinase 1; ASK; ASK 1; ASK1; ASK-1; EC 2.7.11.25; kinase ASK1; M3K5; MAP/ERK kinase kinase 5; Map3k5; MAPK/ERK kinase kinase 5; MAPKKK5; MEK kinase 5; MEKK 5; Mekk5; mitogen activated protein kinase kinase kinase 5; mitogen-activated protein kinase kinase kinase 5; RGD1306565; RP3-325F22.4
Common Name ASK1
Gene Symbol MAP3K5
KinaseFamily STE11 Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
Kinase Group Ser⁄Thr Kinases: STE Group
Shipping Condition Dry Ice
System Compatibility LanthaScreen™ Antibody, LanthaScreen™ Substrate
Target Entry MAP3K5
Target Gene mitogen-activated protein kinase kinase kinase 5
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.