Learn More
Invitrogen™ Human MAP3K3 (MEKK3), GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length
Specifications
Specifications
| Accession Number | Q99759 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 4215 |
| Molecular Weight (g/mol) | 99.8 kDa |
| Name | Human MAP3K3 (MEKK3), GST Tag |
| Quantity | 100 μg |
| Regulatory Status | RUO - research use only |
| Gene Alias | AW548911; MAP/ERK kinase kinase 3; MAP3K3; MAPK/ERK kinase kinase 3; MAPKKK3; MEK kinase 3; MEKK 3; Mekk3; mitogen activated protein kinase kinase kinase 3; mitogen-activated protein kinase kinase kinase 3; mKIAA4031 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.