missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAP3K15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103334
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MAP3K15.
MAP3K15 is a component of a protein kinase signal transduction cascade.
Specifications
| MAP3K15 | |
| Polyclonal | |
| Unconjugated | |
| MAP3K15 | |
| apoptosis signal-regulating kinase 3; ASK3; bA723P2.3; BC031147; map3k15; MAPK/ERK kinase kinase 15; MEK kinase 15; MEKK 15; MEKK15; mitogen-activated protein kinase kinase kinase 15 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 270672, 389840 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A2AQW0, Q6ZN16 | |
| MAP3K15 | |
| A synthesized peptide derived from human MAP3K15(Accession Q6ZN16), corresponding to amino acid residues E1210-K1260. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction