missing translation for 'onlineSavingsMsg'
Learn More

MAP3K12 binding inhibitory protein 1 Antibody (2E2), Novus Biologicals™

Product Code. 18337509 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18337509 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18337509 Supplier Novus Biologicals Supplier No. H00051562M05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MAP3K12 binding inhibitory protein 1 Monoclonal antibody specifically detects MAP3K12 binding inhibitory protein 1 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP3K12 binding inhibitory protein 1
Applications ELISA, Immunocytochemistry, Sandwich ELISA
Classification Monoclonal
Clone 2E2
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH05197
Gene Alias MAP3K12 binding inhibitory protein 1, MAP3K12-binding inhibitory protein 1, MAPK upstream kinase-binding inhibitory protein, MUK-binding inhibitory protein
Host Species Mouse
Immunogen MBIP (AAH05197, 1 a.a. ~ 344 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAAATEHNRPSSGDRNLERRCSPNLSREVLYEIFRSLHTLVGQLDLRDDVVKITIDWNKLQSLSAFQPALLFSALEQHILYLQPFLAKLQSPIKEENTTAVEETGRTEMGNKNEVNDKFSIGDLQEEEKHKESDLRDVKKTQIHFDPEVVQIKAGKAEIDRRISAFIERKQAEINENNVREFCNVIDCNQENSCARTDAIFTPYPGFKSHVKVSRVVYTYGPQTRPEGIPGSGHKPNSMLRDCGNQAVEERLQNIEAHLRLQTGGPVPRDIYQRIKKLEDKILELEGISPEYFQSVSFSGKRRKVQPPQQNYSLAELDEKISALKQALLRKSREAESMATHHLP
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 51562
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.