missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MAP3K12 Polyclonal specifically detects MAP3K12 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | MAP3K12 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Accession No. | Q12852 |
| Gene Alias | DLKEC 2.7.11.25, Dual leucine zipper bearing kinase, EC 2.7.11, Leucine-zipper protein kinase, MAPK-upstream kinase, MEKK12, mitogen-activated protein kinase kinase kinase 12, Mixed lineage kinase, MUKdual leucine zipper kinase DLK, protein kinase MUK, ZPKleucine zipper protein kinase, ZPKP1 |
| Gene Symbols | MAP3K12 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?