missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP3K12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NBP1-46063-0.1mg
This item is not returnable.
View return policy
Description
MAP3K12 Polyclonal specifically detects MAP3K12 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
Specifications
| MAP3K12 | |
| Polyclonal | |
| Unconjugated | |
| Tris/Glycine buffer pH 7.5-7.9, stabilizing protein, and glycerol with 0.02% Sodium Azide | |
| DLKEC 2.7.11.25, Dual leucine zipper bearing kinase, EC 2.7.11, Leucine-zipper protein kinase, MAPK-upstream kinase, MEKK12, mitogen-activated protein kinase kinase kinase 12, Mixed lineage kinase, MUKdual leucine zipper kinase DLK, protein kinase MUK, ZPKleucine zipper protein kinase, ZPKP1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specifically detects a unique epitope on MAP3K12. | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Immunoprecipitation, Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1:500, ELISA 1:1000, Immunoprecipitation 1:200 | |
| Q12852 | |
| MAP3K12 | |
| Synthetic peptides corresponding to unique epitope on MAP3K12. The peptide sequence was selected from mid-region, amimo acids 270-320. This peptide was covalently modified post-synthetically to achieve desired antigenicity. | |
| 0.1 mg | |
| Protein Kinase | |
| 7786 | |
| Rat, Mouse, Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction