missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MAP3K11 (MLK3), GST Tag Recombinant Protein

Human MAP3K11 (MLK3), GST Tag Recombinant Protein

Brand:  Invitrogen™ PR6962A

Code : NEW

Additional Details : Weight : 0.00900kg

 View more versions of this product

Product Code. 12634770

  • £1602.00 / 100µg
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MAP3K11 (MLK3) activates both the p38 MAP kinase and c-Jun kinase pathways.

Specifications

Specifications

NP_002410.1
120.1 kDa
Human
MLK Kinase Family
Recombinant
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Dry Ice
Kinases & Inhibitors
Liquid
Immunology, Oncology, Signal Transduction
Kinase Assay
100μg
MAP3K11 (MLK3)
Human
GST Tag
Ser⁄Thr Kinases: TKL Group
Baculovirus
Recombinant, Full Length
Serine/Threonine Kinases
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers