missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MAP3K11 (MLK3), GST Tag Recombinant Protein
Human MAP3K11 (MLK3), GST Tag Recombinant Protein
Brand: Invitrogen™ PV4009
Code : NEW
Additional Details : Weight : 0.00900kg
Description
MAP3K11 (MLK3) activates both the p38 MAP kinase and c-Jun kinase pathways.
Specifications
NP_002410.1 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
MAP3K11 (MLK3) | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
120.1kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
MLK Kinase Family | |
Recombinant | |
Baculovirus |