Learn More
MAP3K1 Mouse anti-Human, Clone: 2F6, Abnova™
Mouse monoclonal antibody raised against partial recombinant human MAP3K1.
Brand: Abnova MAB13348.100ug
Description
MAP3K, or MEK kinase, is a serine/threonine kinase that occupies a pivotal role in a network of phosphorylating enzymes integrating cellular responses to a number of mitogenic and metabolic stimuli, including insulin (MIM 176730) and many growth factors.[supplied by OMIM]
Sequence: SKNSMTLDLNSSSKCDDSFGCSSNSSNAVIPSDETVFTPVEEKCRLDVNTELNSSIEDLLEASMPSSDTTVTFKSEVAVLSPEKAENDDTYKDDVNHNQKSpecifications
| MAP3K1 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determ | |
| MAPKKK1/MEKK/MEKK1 | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 4214 | |
| Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| 2F6 | |
| Mouse monoclonal antibody raised against partial recombinant human MAP3K1. | |
| In 10mM PBS. | |
| MAP3K1 | |
| Recombinant protein corresponding to amino acids 1077-1176 of human MAP3K1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.