Learn More
Invitrogen™ Human MAP2K5 (MEK5), GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This MAP2K5 (MEK5) recombinant human protein (full length) was expressed in insect cells. MAP2K5 (MEK5) recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the LanthaScreen™ Eu Kinase Binding assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active The gene accession number for MAP2K5 (MEK5) is AAH08838.1.
Specifications
Specifications
| Accession Number | Q13163 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50mM tris HCl with 0.1mM PMSF, 25% glycerol, 0.25mM DTT, 150mM NaCl, 0.1mM EDTA, 10mM glutathione and no preservative; pH 7.5 |
| Gene ID (Entrez) | 5607 |
| Molecular Weight (g/mol) | 76.4 kDa |
| Name | Human MAP2K5 (MEK5), GST Tag |
| pH Range | 7.5 |
| Purification Method | Purified |
| Quantity | 1 mg |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.