missing translation for 'onlineSavingsMsg'
Learn More

MAP2K3 Rabbit anti-Human, Mouse, Rat, CoraLite 555, Clone: 3O10, Proteintech

Rabbit Recombinant Antibody

Brand:  Proteintech CL555-80137-100UL

 View more versions of this product

Product Code. 17639916

  • £375.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MAP2K3 antibody apecifically detetects MAP2K3 antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence.
Specifications

Specifications

MAP2K3
3O10
CoraLite 555
BC032478
Rabbit
Protein A purification
Primary
Human, Mouse, Rat
Liquid
Immunofluorescence
1000 μg/mL
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
MAP2K3
Peptide
100 μL
5606
Store at -20°C. Avoid exposure to light.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.