missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Human MAP2K1 (MEK1), His Tag Recombinant Protein
Human MAP2K1 (MEK1), His Tag Recombinant Protein
Brand: Thermo Scientific™ PV4003
Code : NEW
Additional Details : Weight : 0.00900kg
Description
- MAP2K1 (MEK1) kinase is a dual-specificity protein kinase
- It functions in mitogen-activated protein kinase (MAPK) cascades, controlling cell growth and differentiation
- MAP2K1 is activated by a wide variety of growth factors and cytokines, and also by membrane-dependent depolarization and calcium influx
Specifications
NP_002746 | |
49.2 kDa | |
MAP2K1 (MEK1) | |
Human | |
His Tag | |
Ser/Thr Kinases: STE Group | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |
Kinase Assay | |
1 mg | |
STE7 Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris pH 7.5, 150mM NaCl, 0.05mM EDTA, 0.05% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid | |
Drug Discovery: Signal Transduction |