missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119846
This item is not returnable.
View return policy
Description
Positive Control: Rat brain tissue lysates, SH-SY5Y, human prostate carcinoma tissue, mouse brain tissue. Subcellular Location: Cytoskeleton, dendrite.
MAP2 (Microtubule Associated Protein 2) exists in two high molecular weight forms (MAP2a & MAP2b) and a low molecular weight form. The expression of MAP2 is developmentally regulated and its multiple forms arise by alternative splicing of a single gene. MAP2 is involved in microtubule assembly, which is an essential step in neurogenesis. The products of similar genes in rat and mouse are neuron-specific cytoskeletal proteins that are enriched in dendrites, implicating a role in determining and stabilizing dendritic shape during neuron development. MAP2 is useful in studies of neuron structure in normal and malignant brain tissue as well as in degenerative diseases such as Alzheimer's Disease (AD).
Specifications
| MAP2 | |
| Polyclonal | |
| Unconjugated | |
| Map2 | |
| a730034c02; AC091465.1; DKFZp686I2148; G1-397-34; MAP 2; Map2; MAP-2; map2.S; map2a; map-2a; map2ab; MAP2B; MAP2C; MAP2R; microtubule associated protein 2; microtubule associated protein 2 S homeolog; microtubule associated protein 2; microtubule-associated protein 2; microtubule associated protein 2C; microtubule-associated protein 2; microtubule-associated protein-2; Mtap2; Mtap-2; OTTHUMP00000163916; OTTHUMP00000208143; repro4; XELAEV_18047318mg; xmap2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17756, 25595, 4133 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| P11137, P15146, P20357 | |
| Map2 | |
| Synthetic peptide within human MAP2 aa 1760-1810. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction