missing translation for 'onlineSavingsMsg'
Learn More

MAP2 Mouse, Clone: AP20, eBioscience™

Product Code. 15517656
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15517656 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15517656 Supplier Invitrogen Supplier No. 14976382

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The monoclonal antibody AP20 recognizes the high molecular weight isoforms of microtubule-associated protein-2 (MAP2a and MAP2b) of human, rat, mouse, bovine, chicken, and xenopus. MAP2 belongs to a family of heat-stable phosphoproteins that are differentially expressed during brain development. MAP2 expression is localized to the cell body and dendrites of neurons and is involved in stabilizing microtubules by crosslinking with intermediate filaments. Expression of MAP2 is commonly used as a neural marker given that its expression becomes more pronounced with neuronal maturation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP2
Applications Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunomicroscopy
Classification Monoclonal
Clone AP20
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Gene Alias Microtubule-associated protein 2
Host Species Mouse
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bovine, Chicken, Human, Mouse, Rat, Xenopus species
Content And Storage Store at 2-8°C.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.