Learn More
Invitrogen™ MAP2 Monoclonal Antibody (MT-08)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA119426
Product Code. 11554662
  • £318.00 / 100µg
Description
This antibody recognizes an epitope (aa 1375-1395) located in central domain of molecule Microtubule Associated Protein 2ab (MAP2ab), an intracellular antigen.
MAP2 (Microtubule Associated Protein 2) exists in two high molecular weight forms (MAP2a & MAP2b) and a low molecular weight form. The expression of MAP2 is developmentally regulated and its multiple forms arise by alternative splicing of a single gene. MAP2 is involved in microtubule assembly, which is an essential step in neurogenesis. The products of similar genes in rat and mouse are neuron-specific cytoskeletal proteins that are enriched in dendrites, implicating a role in determining and stabilizing dendritic shape during neuron development. MAP2 is useful in studies of neuron structure in normal and malignant brain tissue as well as in degenerative diseases such as Alzheimer's Disease (AD).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MAP2
Monoclonal
1 mg/mL
PBS with 15mM sodium azide; pH 7.4
P11137, P20357
Map2
Microtubule protein (bovine brain) enriched for kinesin.
100 μg
Primary
Human, Mouse, Pig, Pig
Antibody
IgG1
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
MT-08
Unconjugated
Map2
a730034c02; AC091465.1; DKFZp686I2148; G1-397-34; MAP 2; Map2; MAP-2; map2.S; map2a; map-2a; map2ab; MAP2B; MAP2C; MAP2R; microtubule associated protein 2; microtubule associated protein 2 S homeolog; microtubule associated protein 2; microtubule-associated protein 2; microtubule associated protein 2C; microtubule-associated protein 2; microtubule-associated protein-2; Mtap2; Mtap-2; OTTHUMP00000163916; OTTHUMP00000208143; repro4; XELAEV_18047318mg; xmap2
Mouse
Protein A
RUO
100153306, 17756, 4133
4°C, do not freeze