missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP2 Antibody (MT-01), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-415DL594
This item is not returnable.
View return policy
Description
MAP2 Monoclonal antibody specifically detects MAP2 in Human, Mouse, Porcine, Bovine, Feline samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| MAP2 | |
| Monoclonal | |
| DyLight 594 | |
| DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, Microtubule Associated Protein 2, microtubule-associated protein 2, MTAP2 | |
| MAP2 Antibody (MT-01) was developed against the Thermostabile fraction of bovine brain microtubules (whole MAP2ab molecule). | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, MAP Kinase Signaling, Neuroscience, Stem Cell Markers | |
| 4133 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| MT-01 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Pig, Bovine, Feline | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction