missing translation for 'onlineSavingsMsg'
Learn More

MAP2 Antibody (4H5), Biotin, Novus Biologicals™

Product Code. 18413682 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18413682 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18413682 Supplier Novus Biologicals Supplier No. NBP225156B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MAP2 Monoclonal antibody specifically detects MAP2 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry Free-Floating
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Free Floating)
Classification Monoclonal
Clone 4H5
Conjugate Biotin
Formulation PBS
Gene Alias DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, Microtubule Associated Protein 2, microtubule-associated protein 2, MTAP2
Host Species Mouse
Immunogen MAP2 Antibody (4H5) was developed against full length purified bovine protein, epitope mapped to projection domain of human sequence, between amino acids 631 and 1056.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, MAP Kinase Signaling, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4133
Target Species Human, Mouse, Rat, Bovine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.