missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAP1S Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562995
This item is not returnable.
View return policy
Description
Immunogen sequence: RCGEASPPSA ACGSPASQLV ATPSLELGPI PAGEEKALEL PLAASSIPRP RTPSPESHRS PAEGSERLSL SPL Highest antigen sequence identity to the following orthologs: Mouse - 35%, Rat - 32%.
Microtubule-associated protein that mediates aggregation of mitochondria resulting in cell death and genomic destruction (MAGD). Plays a role in anchoring the microtubule organizing center to the centrosomes. Binds to DNA. Plays a role in apoptosis. Involved in the formation of microtubule bundles.
Specifications
| MAP1S | |
| Polyclonal | |
| Unconjugated | |
| MAP1S | |
| BPY2 interacting protein 1; BPY2-interacting protein 1; BPY2IP1; C19orf5; MAP1S; MAP-1S; MAP1S heavy chain; MAP1S light chain; MAP8; microtubule associated protein 1S; microtubule-associated protein 1S; Microtubule-associated protein 8; variable charge Y chromosome 2-interacting protein 1; VCY2 interacting protein 1; VCY2-interacting protein 1; VCY2IP1; VCY2IP-1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55201 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q66K74 | |
| MAP1S | |
| Recombinant protein corresponding to Human MAP1S. Recombinant protein control fragment (Product #RP-101274). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction