missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP1S Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-47282AF700
This item is not returnable.
View return policy
Description
MAP1S Polyclonal antibody specifically detects MAP1S in Human samples. It is validated for Western Blot
Specifications
| MAP1S | |
| Polyclonal | |
| Western Blot | |
| BPY2 interacting protein 1, BPY2IP1VCY2IP-1, C19orf5, chromosome 19 open reading frame 5, FLJ10669, MAP8MAP-1S, MGC133087, microtubule-associated protein 1S, Microtubule-associated protein 8, Variable charge Y chromosome 2-interacting protein 1, VCY2 interacting protein 1, VCY2-interacting protein 1, VCY2IP1BPY2-interacting protein 1 | |
| The immunogen for this product maps to a region between residue 525 and 575 of human MAP1S using the numbering given in entry NP_060644.4 (GeneID 55201). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 55201 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction