Learn More
Invitrogen™ MAP1A Monoclonal Antibody (HM-1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA532988
Description
Reconstitute by adding 1 mL of PBS buffer to yield a concentration of 100 μg/mL. Positive Control - IHC: Rat Brain Tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Structural protein involved in the filamentous cross-bridging between microtubules and other skeletal elements.
Specifications
| MAP1A | |
| Monoclonal | |
| 100 μg/mL | |
| Ascites, 1.2% sodium acetate with 2mg BSA and 0.01mg sodium azide | |
| P34926, Q9QYR6 | |
| MAP1A | |
| Rat brain microtubule-associated proteins (MAPs). | |
| RUO | |
| 17754, 25152 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| HM-1 | |
| Unconjugated | |
| MAP1A | |
| 6330416M19Rik; AI853608; MAP1 light chain LC2; MAP1A; MAP-1A; MAP1A heavy chain; MAP1L; microtubule associated protein 1A; microtubule-associated protein 1 A; microtubule-associated protein 1A; modifier of tubby hearing 1; moth1; Mtap1; Mtap-1; Mtap1a; Proliferation-related protein p80 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.