missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP1 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97771PCP
This item is not returnable.
View return policy
Description
MAP1 Polyclonal antibody specifically detects MAP1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| MAP1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry-Paraffin | |
| MAP1, MAP-1PNMA4Paraneoplastic antigen Ma4, modulator of apoptosis 1, paraneoplastic antigen like 4 | |
| Produced in rabbits immunized with E. coli-derived Human MAP1 fragment. | |
| 0.1 mL | |
| Apoptosis, MAP Kinase Signaling | |
| 64112 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction