missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76453-0.025mg
This item is not returnable.
View return policy
Description
MAP1 Polyclonal specifically detects MAP1 in Human, Mouse samples. It is validated for Western Blot, ELISA.
Specifications
| MAP1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| MAP1, MAP-1PNMA4Paraneoplastic antigen Ma4, modulator of apoptosis 1, paraneoplastic antigen like 4 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1-4 μg/mL, ELISA 1:100-1:2000 | |
| NP_071434 | |
| MOAP1 | |
| Antibody was raised against a 15 amino acid synthetic peptide from near the center of human MAP-1. The immunogen is located within the last 50 amino acids of MAP1. Amino Acid Squence: CKDYEAAEEEEALLQA | |
| 0.025 mg | |
| Apoptosis, MAP Kinase Signaling | |
| 64112 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction