Learn More
MAP 2a,b,c (Microtubule-Associated Protein) Ab-3 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry experiments with the Epredia™ MAP 2a,b,c (Microtubule-Associated Protein) Ab-3, Mouse Monoclonal Antibody.
Brand: Epredia MS-250-R7
Description
Microtubule Associated Protein 2 exists in two high molecular weight forms (2a & 2b) and a low molecular weight form. The expression of MAP2 is developmentally regulated and its multiple forms arise by alternative splicing of a single gene. Ab-3 is specific to phosphprylated form of MAP2a,b,c and shows no cross-reaction with other MAPs, tau, and tubulin. Ab-3 reacts with dendrites and cell bodies of neurons. It is useful in studies of neuron structure in normal and malignant brain tissue and in Alzheimer's.Host Species: Mouse
Clone: AP18
Isotype: IgG1
Species Reactivity: Human, Cow, Rat, Xenopus, Quail. Others not known.
Epitope: aa 1-151
Immunogen: Bovine brain microtubule associated protein 2
Molecular Weight: ∽280kDa doublet and a 70kDa singlet
Positive Control: IMR-5 or T98G human glioblastoma cells or brain
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
Specifications
| MAP 2a,b,c (Microtubule-Associated Protein) Ab-3 | |
| Monoclonal | |
| — | |
| Mouse | |
| 7 mL | |
| Neuroscience | |
| Bovine, Human, Quail, Rat, Xenopus species | |
| IgG1 |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| AP18 | |
| Unconjugated | |
| Bovine brain microtubule associated protein 2 | |
| IVD | |
| Primary | |
| Prediluted |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.