missing translation for 'onlineSavingsMsg'
Learn More

MAP 2a,b,c (Microtubule-Associated Protein) Ab-3 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry experiments with the Epredia™ MAP 2a,b,c (Microtubule-Associated Protein) Ab-3, Mouse Monoclonal Antibody.

Brand:  Epredia MS-250-PABX

 View more versions of this product

Product Code. 12620707

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Microtubule Associated Protein 2 exists in two high molecular weight forms (2a & 2b) and a low molecular weight form. The expression of MAP2 is developmentally regulated and its multiple forms arise by alternative splicing of a single gene. Ab-3 is specific to phosphprylated form of MAP2a,b,c and shows no cross-reaction with other MAPs, tau, and tubulin. Ab-3 reacts with dendrites and cell bodies of neurons. It is useful in studies of neuron structure in normal and malignant brain tissue and in Alzheimer's.

Host Species: Mouse

Clone: AP18

Isotype: IgG1

Species Reactivity: Human, Cow, Rat, Xenopus, Quail. Others not known.

Epitope: aa 1-151

Immunogen: Bovine brain microtubule associated protein 2

Molecular Weight: ∽280kDa doublet and a 70kDa singlet

Positive Control: IMR-5 or T98G human glioblastoma cells or brain

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MAP 2a,b,c (Microtubule-Associated Protein) Ab-3
Monoclonal
1mg/mL
Mouse
200 μL
Neuroscience
Bovine, Human, Quail, Rat, Xenopus species
IgG1
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
AP18
Unconjugated
Bovine brain microtubule associated protein 2
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.