missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAOB Mouse anti-Human, Clone: 2B12F3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66107-1-IG-20UL
This item is not returnable.
View return policy
Description
The MAOB gene encodes a 520-amino acid protein with a molecular mass of 58.8 kDa and shows 70% amino acid identity to MAOA. MAOA and MAOB are present in the outer mitochondrial membrane in the central nervous system and peripheral tissues. A comparison of highly purified human placental MAOA and human liver MAOB revealed that the A form of the enzyme is larger by 2 kDa and only one potential N-glycosylation site exists in each protein, with each site in a different relevant position.Specifications
| MAOB | |
| Monoclonal | |
| 0.73 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P27338 | |
| MAOB | |
| MAOB Fusion Protein Ag17918 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2B12F3 | |
| Unconjugated | |
| MAOB | |
| MAO B, MAOB, monoamine oxidase B, Monoamine oxidase type B | |
| Mouse | |
| Protein G | |
| RUO | |
| 4129 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction