missing translation for 'onlineSavingsMsg'
Learn More

MAOB Mouse anti-Human, Clone: 2B12F3, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66107-1-IG-150UL

 View more versions of this product

Product Code. 16817685

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The MAOB gene encodes a 520-amino acid protein with a molecular mass of 58.8 kDa and shows 70% amino acid identity to MAOA. MAOA and MAOB are present in the outer mitochondrial membrane in the central nervous system and peripheral tissues. A comparison of highly purified human placental MAOA and human liver MAOB revealed that the A form of the enzyme is larger by 2 kDa and only one potential N-glycosylation site exists in each protein, with each site in a different relevant position.
Specifications

Specifications

MAOB
Monoclonal
0.73 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P27338
MAOB
MAOB Fusion Protein Ag17918
150 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
2B12F3
Unconjugated
MAOB
MAO B, MAOB, monoamine oxidase B, Monoamine oxidase type B
Mouse
Protein G
RUO
4129
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.