missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MAOA Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA586581

Product Code. 16373455

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Purity is > 95% by SDS-PAGE.

This gene encodes monoamine oxidase A, an enzyme that degrades amine neurotransmitters, such as dopamine, norepinephrine, and serotonin. The protein localizes to the mitochondrial outer membrane. The gene is adjacent to a related gene on the opposite strand of chromosome X. Mutation in this gene results in monoamine oxidase deficiency, or Brunner syndrome.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MAOA
Polyclonal
Unconjugated
MAOA
1110061B18Rik; AA407771; Amine oxidase [flavin-containing] A; HGNC:6833; Mao; MAOA; MAO-A; monoamine oxidase A; monoamine oxidase type A; RP1-201D17__B.2
Rabbit
Antigen affinity chromatography
RUO
17161, 29253, 4128
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
P21396, P21397, Q64133
MAOA
Synthetic peptide, corresponding to amino acids 330-380 of Human MAOA.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.