missing translation for 'onlineSavingsMsg'
Learn More

MAO-IN-1, MedChemExpress

Product Code. 16473089
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473089 1 mg 1mg
16470109 10 mg 10mg
16422308 5 mg 5mg
16400636 20 mg 20mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16473089 Supplier Medchem Express Supplier No. HYU00015
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress MAO-IN-1 is a monoamine oxidase B (MAO B) inhibitor with an IC50 of 20 nM.

MAO-IN-1 is a monoamine oxidase B (MAO B) inhibitor with an IC50 of 20 nM.

Specifications

Chemical Name or Material MAO-IN-1
CAS 124991-40-8
Color White
Purity Grade Notes Research
Molecular Formula C17H19ClO4
Quantity 1 mg
SMILES O[C@@H](COC)COC1=CC=C(OCC2=CC=CC(Cl)=C2)C=C1
Molecular Weight (g/mol) 322.78
Formula Weight 322.78
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.