missing translation for 'onlineSavingsMsg'
Learn More

Manumycin A, MP Biomedicals™

Product Code. 15907315 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15907315 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15907315 Supplier MP Biomedicals Supplier No. 0218366605

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

An antibiotic that act as a potent and selective farnesyltransferase inhibitor

  • By adding a farnesyl group to the -SH of the cysteine near the end of target proteins, Farnesyltransferase posttranslationally-modifies proteins to form a thioether linkages
  • Used as an antibiotic and different research branches
  • Farnesylation enables the proteins to become membrane associated
TRUSTED_SUSTAINABILITY

Specifications

CAS 52665-74-4
Synonym Manumycin A, manumycin, Ucfi-C, AC1O5PHE, UNII-OIQ298X4XD
Quantity 5 mg
Molecular Formula C31H38N2O7
Color Brown-Orange
Purity >95
Molecular Weight (g/mol) 550.65
Chemical Name or Material Manumycin A
compliance-icons compliance-icons
Product Identifier
  • Primaquine bisphosphate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H341-Suspected of causing genetic defects.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
  • MIXTURE LIST-Contain: Primaquine bisphosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.