missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Sturdy cover closes over troughs to minimize evaporation.
- Baskets are designed to lock slides into position even when turned upside down.
- Slide baskets hold 20 standard microscope slides.
- Troughs hold 11 fl. oz. (320mL) of reagent.
- Best when used for low-volume procedures.
Physical Specifications
Dimensions (D x W x H): 7.1 x 39.4 x 4.7″ (18 x 100 x 12cm)
Includes:
Kit includes a steel support rack, a complete set of 14 staining troughs (320mL, No. A78010487) and five slide baskets with carriers (No. A78010466).
Specifications
Specifications
| Slide Capacity | 20 |
| Reagent Capacity | 320 mL |
| Dimensions (L x W x H) | 7.1 x 39.4 x 4.7 in. (18 x 100 x 12 cm) |
| Height (English) | 4.7 in. |
| Height (Metric) | 12 cm |
| Length (English) | 7.1 in. |
| Length (Metric) | 18 cm |
| Width (English) | 39.4 in. |
| Width (Metric) | 100 cm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?