Learn More
Specac™ Manual Hydraulic Conversion Press Kit 0-5 Ton
Description
0-5 Ton Conversion Kit for use with Manual Press GS15011. This kit provides an additional load gauge to be used along with the standard 15 Ton load gauge on the Press for more accurate reading at lower loads.
Specifications
Specifications
| For Use With (Equipment) | Manual Press GS15011 |