Learn More
Specac™ Manual Hydraulic Conversion Press Kit 0-2 Ton
Product Code. 15230168
Encompass_Preferred
Click to view available options
For Use With (Equipment):
Manual Press GS15011
Product Code. 15230168
Brand: Specac™ GS15052
0-2 Ton Conversion Kit for use with Manual Press GS15011. This kit provides an additional load gauge to be used along with the standard 15 Ton load gauge on the Press for more accurate reading at lower loads.
Specifications
For Use With (Equipment) Manual Press GS15011