Learn More
Specac™ Manual Hydraulic Conversion Press Kit 0-1 Ton
Product Code. 15220168
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Manual Press GS15011
Product Code. For Use With (Equipment)
15220168 Manual Press GS15011
1 options
Product Code. 15220168 Supplier Specac™ Supplier No. GS15051
Estimated Shipment: 22-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
0-1 Ton Conversion Kit for use with Manual Press GS15011. This kit provides an additional load gauge to be used along with the standard 15 Ton load gauge on the Press for more accurate reading at lower loads.
Specifications
For Use With (Equipment) Manual Press GS15011