Learn More
- Home
- Products
- Vials
- Vial Capping and Decapping Equipment
- Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers
Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers
Product Code. 10301535| Product Code. | Size | Type | unitSize |
|---|---|---|---|
| 10301535 | 30 mm | Manual Decrimper | Each |
| 15280389 | 8 mm | Manual Crimper | Each |
| 14860742 | 11 mm | Manual Crimper | Each |
| 13488988 | 20 mm | Manual Crimper | Each |
| 10147664 | 8 mm | Manual Crimper | Each |
| 10429454 | 11 mm | Manual Crimper | Each |
| 10566765 | 13 mm | Manual Crimper | Each |
| 10231585 | 13 mm | Manual Crimper (Flip-off) | Each |
| 12400268 | 20 mm | Manual Crimper | Each |
| 10372525 | 20 mm | Manual Crimper (Flip-off) | Each |
| 10250535 | 28 mm | Manual Crimper | Each |
| 10773667 | 30 mm | Manual Crimper | Each |
| 10229964 | 32 mm | Manual Crimper | Each |
| 11528470 | 11 mm | Manual Crimper | Each |
| 11538470 | 20 mm | Manual Crimper | Each |
| 15290389 | 8 mm | - | Each |
| 15188784 | 20 mm | - | Each |
| 10259614 | 8 mm | Manual Decrimper | Each |
| 10488084 | 11 mm | Manual Decrimper | Each |
| 10240535 | 13 mm | Manual Decrimper | Each |
| 10279364 | 20 mm | Manual Decrimper | Each |
| 12852344 | 28 mm | Manual Decrimper | Each |
| 11528240 | 32 mm | Manual Decrimper | Each |
| 11564815 | 20 mm | Manual Crimper | Each |
| 10530303 | 11 mm | Manual Decrimper | Each |
| 10143362 | 20 mm | Manual Decrimper | Each |
| 12054907 | 11 mm | Manual Decapping Plier | Each |
| 10299454 | 13 mm | Manual Decapping Plier | Each |
| 10066164 | 20 mm | Manual Decapping Plier | Each |
| 10200205 | 30 mm | Manual Decapping Plier | Each |
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Description
Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers, also for use with National or Chromacol vials.
- Minimizes damage to vial rim with chromatography or serum vials
- Removes standard aluminum seals and many steel seals
Specifications
Specifications
| Description | Manual Decrimper |
| No. per Pack | 1 Ea. |
| Type | Manual Decrimper |
| For Use With (Equipment) | Aluminum And Steel Seal, National Vial, Chromacol Vial |
| Size | 30 mm |
| For Use With (Application) | Industrial Chromatography |
| Product Line | National |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.