Learn More
Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers
Product Code. 12852344
Change view
Click to view available options
Size:
8 mm
11 mm
13 mm
20 mm
28 mm
30 mm
32 mm
Type:
Manual Crimper
Manual Crimper (Flip-off)
Manual Decapping Plier
Manual Decrimper
Unit Size:
Each
30 product options available for selection
Product selection table with 30 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Type unitSize
12852344 28 mm Manual Decrimper Each
15280389 8 mm Manual Crimper Each
14860742 11 mm Manual Crimper Each
13488988 20 mm Manual Crimper Each
10147664 8 mm Manual Crimper Each
10429454 11 mm Manual Crimper Each
10566765 13 mm Manual Crimper Each
10231585 13 mm Manual Crimper (Flip-off) Each
12400268 20 mm Manual Crimper Each
10372525 20 mm Manual Crimper (Flip-off) Each
10250535 28 mm Manual Crimper Each
10773667 30 mm Manual Crimper Each
10229964 32 mm Manual Crimper Each
11528470 11 mm Manual Crimper Each
11538470 20 mm Manual Crimper Each
15290389 8 mm - Each
15188784 20 mm - Each
10259614 8 mm Manual Decrimper Each
10488084 11 mm Manual Decrimper Each
10240535 13 mm Manual Decrimper Each
10279364 20 mm Manual Decrimper Each
10301535 30 mm Manual Decrimper Each
11528240 32 mm Manual Decrimper Each
11564815 20 mm Manual Crimper Each
10530303 11 mm Manual Decrimper Each
10143362 20 mm Manual Decrimper Each
12054907 11 mm Manual Decapping Plier Each
10299454 13 mm Manual Decapping Plier Each
10066164 20 mm Manual Decapping Plier Each
10200205 30 mm Manual Decapping Plier Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 30 options available.
30 options
Product Code. 12852344 Supplier Thermo Scientific™ Supplier No. C4028102
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Achieve secure, repeatable crimps or remove seals with minimized risk of damage.
Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers, also for use with National or Chromacol vials.
Place decrimping head squarely on seal and squeeze.
  • Minimizes damage to vial rim with chromatography or serum vials 
  • Removes standard aluminum seals and many steel seals
TRUSTED_SUSTAINABILITY
Specifications
Description Manual Decrimper
No. per Pack 1 Ea.
Type Manual Decrimper
For Use With (Equipment) Aluminum And Steel Seal, National Vial, Chromacol Vial
Size 28 mm
For Use With (Application) Industrial Chromatography
Product Line National
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.