Learn More
Sartorius Manual Cubis™ MCM Mass Comparator with Climate Module
Product Code. 16470114 Shop All Sartorius Products
Change view
Click to view available options
Type:
Mass Comparator
Unit Size:
Each
Product Code. Type Unit Size
16470114 Mass Comparator Each
1 options
Product Code. 16470114 Supplier Sartorius Supplier No. MCM605
  • ONLINE EXCLUSIVE PRICE
    £33560.00
    £32970.00 / Pack of 1
    Save £590.00
    1% Off
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Manual Cubis™ MCM Mass Comparator with Climate Module combines metrological expertise and integrated OIML R 111-1 workflows. A choice of 14 models is available with maximum capacities of 6 g to 64 kg and readabilities of 0.1 μg to 10 mg.
  • Complete mass standard laboratory in a single unit
  • Integrated climate sensors for recording all data relevant for determining measurement uncertainty
  • Integrated workflow control for efficient and error-free mass comparison
  • Fast measurement cycles according to the ABA, ABBA, AB1...BnA
  • Cubis™ MSA color touch screen for fast and simple configuration of parameters and workflows
  • Sensor-equipped climate module integrated into the draft shield for recording the temperature, humidity and air pressure
  • Integrated calibration workflows for ABA, ABBA, AB1..BnA cycles to ensure efficient, error-free mass comparison
  • Fully integrated function for determining the measurement uncertainty in accordance with OIML and ASTM recommendations
  • Filters for optimal adaptation of the mass comparator to ambient conditions
  • Monolithic weighing technology
  • For display and evaluation, complete electronics and power supply separated from the weighing system to prevent heat from affecting the results
  • All MCM mass comparators featuring eccentric (off-center) load compensation for easy loading of weights without automatic centering
  • Additional applications for density determination, statistics and individual identifiers are integrated as standard programs
  • Built-in SD card slot for storage and transfer of all data and settings
  • Graphical level indicator for interactive user guidance during levelling
  • Easy logging of reference weight data
  • Continuous weighing range display: any weight between 0 g and the maximum capacity can be displayed
  • USB, RS-232C and Ethernet interface ports to integrate the mass comparator into networks or to enable it to communicate with external software via third-party protocols, standardized communication protocols or Web services
TRUSTED_SUSTAINABILITY
Specifications
Type Mass Comparator